missing translation for 'onlineSavingsMsg'
Learn More

AGMA Sterile Zyceine Disinfectant Wipe

Product Code. 30066733
Change view
Click to view available options
Dimensions (L x W):
28 x 28 cm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
30066733 28 x 28 cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066733 Supplier AGMA Supplier No. 9401
  • ONLINE EXCLUSIVE PRICE
    208.95 €
    189.87 € / Pack of 10
    Save 19.08 €
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect your cleanrooms with a unique two-component sporicide that offers high microbial efficacy once activated.

Totally stable until activated, Zyceine has a proven fungicidal efficacy of greater than a 3-log reduction in 2 minutes, and a yeasticidal efficacy greater than a 4-log reduction in 2 minutes.

This solution has met the MHRA/NHS requirement for a sporicide for their 'wiping in' process which can achieve a 2 log reduction in 2mins. Available in a liquid pouch or high-quality wipes.

  • Fast acting Sporicid
  • Independently validated efficacy
  • WFI quality water – High Purity Water
  • Available with 28 x 38 cm wipes, allowing for large area coverage
  • Also available as 200 mL or 1 L fluid pouch, ideal for using with a mop system
  • Fully traceable with supporting analysis and certification
  • Gamma irradiation for Sterility
  • 2 year unopened helf life
  • 8 hour efficacy once activated
  • EN13704, EN1276, EN1650, EN14348

Specifications

Product Type Disinfectant Wipe
Length (English) 14.96 in.
Length (Metric) 38 cm
Width (English) 11.02 in.
Width (Metric) 28 cm
Material Polycellulose
Autoclavable Not Autoclavable
Container Type Burstable Pouch
Disposable Limited-use
Dimensions (L x W) 28 x 28 cm
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Packaging Quantity 10 x 5 Wipes
Sterility Sterile
Show More Show Less
Product Identifier
  • Sterile Zyceine in WFI High Quality
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Hydroxide, Citric Acid Anhyrdous,Sodium Dichloroisocyanurate Anhydrous

Use biocides safely. Always read the label and product information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.