missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Autoclave Bags
GREENER_CHOICE

Product Code. 11399093
Change view
Click to view available options
Dimensions (L x W):
601 x 406mm
610 x 406mm
660 x 305mm
810 x 610mm
Material:
HDPE
Polypropylene
No. per Case:
200
500
Unit Size:
Pack of 200
Pack of 500
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Material No. per Case Unit Size
11399093 660 x 305mm HDPE 200 Pack of 200
11309103 610 x 406mm HDPE 200 Pack of 200
11359103 660 x 305mm HDPE 500 Pack of 500
11844145 610 x 406mm HDPE 500 Pack of 500
11339103 810 x 610mm HDPE 200 Pack of 200
11379103 660 x 305mm Polypropylene 200 Pack of 200
11349103 660 x 305mm Polypropylene 500 Pack of 500
11389103 610 x 406mm Polypropylene 200 Pack of 200
11369103 601 x 406mm Polypropylene 500 Pack of 500
11329103 810 x 610mm Polypropylene 200 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11399093 Supplier Thermo Scientific™ Supplier No. 50917
  • ONLINE EXCLUSIVE PRICE
    113.53 €
    69.16 € / Pack of 200
    Save 44.37 €
    39% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags

Ordering Alerts:

Not available in all geographic regions

TRUSTED_SUSTAINABILITY

Specifications

Description Autoclave bags (HDPE)
Material HDPE
No. per Case 200
Temperature Range (Metric) 121°C
Dimensions (L x W) 660 x 305mm
Length (English) 26 in.
Width (English) 12 in.
Length (Metric) 660 mm
Width (Metric) 305 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.