missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Disposable Glass Containers

Product Code. p-6223925
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Pack of 375
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12882793 30 mL Pack of 375
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12882793 Supplier Thermo Scientific™ Supplier No. 39404

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for small volume samples, Thermo Scientific™ Sterilin™ Disposable Glass Containers are made from soda glass with a leakfree polypropylene cap.

Ideal for small volume samples, Thermo Scientific™ Sterilin™ Disposable Glass Containers are made from soda glass with a leakfree polypropylene cap.

NOT AVAILABLE IN AMERICA

  • Soda glass offers superior chemical resistance to plastic but is not recommended for autoclaving
  • Tray packed for safety and convenience
  • Supplied with cap—replacement caps available for the 30mL universal
  • Supplied unlabeled and nonsterile
  • In vitro use only

Ordering Information:

  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Material Soda Lime Glass
Sterility Non-sterile
Capacity (Metric) 30 mL
Closure Material Polypropylene
Description Disposable Glass Container
No. per Case 375/Cs.

Note: This glassware is manufactured from Soda glass and therefore it is recommended that the product is not autoclaved.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.