Learn More
Thermo Scientific™ Sterilin™ Loops and Spreaders
Product Code. 11369283
Change view
Click to view available options
Volume:
1 μL
10 μL
Product Type:
Inoculating Loop and Needle, Flexible
Inoculating Loop and Needle, Rigid
Inoculating Loop, Spherical Tip
Inoculating Needle
L-Shaped Spreader
Color:
Blue
Dark blue
Dark green
Green
Pale Green
Violet
Quantity:
10
500
1000
Unit Size:
Pack of 1000
Pack of 500
Pack of 800
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Product Type Color Quantity Unit Size
11369283 10 μL Inoculating Loop and Needle, Rigid Dark blue 10 Pack of 800
11379283 1 μL Inoculating Loop and Needle, Rigid Dark green 10 Pack of 800
11834732 1 μL Inoculating Loop and Needle, Flexible Pale Green 10 Pack of 800
11778858 1 μL Inoculating Loop, Spherical Tip Green 1000 Pack of 1000
11788858 10 μL Inoculating Loop, Spherical Tip Blue 1000 Pack of 1000
11718868 - Inoculating Needle Violet 1000 Pack of 1000
11788978 - L-Shaped Spreader Blue 500 Pack of 500
11728868 - L-Shaped Spreader Blue 1000 Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11369283 Supplier Thermo Scientific™ Supplier No. SL10H
Quantity Price
1 176.79 € / Pack of 800
10 102.47 € / Pack of 800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
NOT AVAILABLE IN AMERICA
  • For inoculation, dilution streaking, spreading and picking isolated colonies
  • Eliminate the need for flaming and the subsequent risk of aerosols Inoculation loop for fixed sample volumes with choice of sphere for dilution streaking, or needle for picking colonies and stab cultures
  • Choice of rigid or flexible loops to suit different applications and user preferences
  • Hexagonal shaft improves grip and assists orientation
  • Free of lubricants, oils and electrostatic charges to facilitate consistent wetting and complete liquid transfer
  • No rough edges means smooth, problem-free plating and streaking of cultures
  • Sterilized by gamma irradiation
Ordering Information
  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifications
Volume 10 μL
Sterility Irradiated
Color Dark blue
Product Type Inoculating Loop and Needle, Rigid
Quantity 10