Learn More
Thermo Scientific™ Sterilin™ Water Sampling Bottles, 500mL PET Square bottle, labeled, dosed 120mg/L with Sodium Thiosulphate
GREENER_CHOICE
Product Code. 13128770
Product Code. 13128770 Supplier Thermo Scientific™ Supplier No. WSC500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Sterilin Water Sampling Bottles.
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Thermo Scientific™ Sterilin Water Sampling Bottles.
  • Available manufactured using:
    • Flexible polystyrene
    • Robust non-toxic PETG
    • NEW, lightweight PRT for improved handling
  • Empty or pre-dosed with sodium thiosulphate
  • Polystyrene and PETG bottles feature color-coded labels to aid differentiation:
    • Blue: dosed
    • Green: undosed
  • Tamper evident for sample integrity
  • Lot number for complete traceability
  • Sterilized by gamma irradiation
  • Dose level 20mg/h: Suitable for neutralizing water samples with a low concentration of chlorine
  • Dose level 120mg/h: Suitable for neutralizing water samples with a high concentration of chlorine