missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ White Caps to use with M00293 Specimen Tubes
GREENER_CHOICE

Product Code. 15311281
Click to view available options
Color:
White
Unit Size:
Pack of 5000
This item is not returnable. View return policy

Product Code. 15311281

Brand: Thermo Scientific Sterilin M00491

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Screw Cap
Color White
Quantity 5000 Pack
For Use With (Equipment) M00293 Specimen Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.