missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Sterilized 1.25 mL Dispenser Tips

Product Code. 10303993
Change view
Click to view available options
Volume (Metric):
1.25 mL
Unit Size:
Pack of 100
Product Code. Volume (Metric) unitSize
10303993 1.25 mL Pack of 100
1 options
This item is not returnable. View return policy

Product Code. 10303993

Brand: Ratiolab™ 2910102

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use in serial pipetting applications.

  • Dispenser tips fit onto Ratiolab™ dispetta and Eppendorf Multipette™ 4780, Ritter Dispenser, Gilson Distriman and Brand HandyStep S
  • Not autoclavable
  • Visible graduations
  • Sterilized
  • Provided in packs of 100

Specifications

Volume (Metric) 1.25 mL
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Format Bulk
Quantity 100/pk
Product Type Dispenser Tips
For Use With (Equipment) Serial pipetting
Graduated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.