missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sterillin™ Silicone Discs
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Used as highly effective cap liners or as a septa for injection ports/pierceable closures and wide variety of applications |
| Material | Rubber |
| Diameter (Metric) | 14 mm |
| Product Type | Disc |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?