missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
- This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged for disposable use and sterilised by gamma rays
- Production, assembly and packaging according to clean room class 7
- Can be detected by colour scanners, metal detectors or X-rays
Compliance
EU food requirements
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Blue |
| Disposable | Single-use |
| Product Type | Detectable Sampling Scoop |
| For Use With (Application) | Sampling Powders, Granulates and Fluids |
| Length (English) | 11.02 in. |
| Material | Polystyrene |
| Length (Metric) | 280 mm |
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
