missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Detectable Scoops
BIOPHARMA

Product Code. 15634114 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
Length (Metric):
141 mm
160 mm
205 mm
216 mm
232 mm
280 mm
332 mm
Unit Size:
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Unit Size
15634114 25 mL 141 mm Pack of 10
15604124 250 mL 232 mm Pack of 10
15614124 500 mL 280 mm Pack of 10
15624124 1000 mL 332 mm Pack of 10
15644114 50 mL 160 mm Pack of 10
15654114 100 mL 205 mm Pack of 10
15664114 150 mL 216 mm Pack of 10
17200594 2000 mL - Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15634114 Supplier Buerkle™ Supplier No. 53782001
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For sampling food, animal feed and pharmaceutical industries. Buerkle™ SteriPlast™ Blue Detectable Scoops are ideal for powders, granulates and pastes and can be detected by color scanners, metal detectors and X rays.

  • Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
  • This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
  • Can be used as part of HACCP/IFS foreign object management
  • Individually packaged for disposable use and sterilised by gamma rays
  • Production, assembly and packaging according to clean room class 7
  • Can be detected by colour scanners, metal detectors or X-rays

Compliance

EU food requirements

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 25 mL
Color Blue
Disposable Single-use
Product Type Detectable Sampling Scoop
For Use With (Application) Sampling Powders, Granulates and Pastes
Length (English) 5.5 in.
Material Polystyrene
Length (Metric) 141 mm
Sterility Sterile
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.