missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Detectable Spoons
BIOPHARMA

Product Code. 15684114 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
2.5 mL
10 mL
Length (Metric):
127 mm
170 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Unit Size
15684114 2.5 mL 127 mm Pack of 100
15694114 10 mL 170 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15684114 Supplier Buerkle™ Supplier No. 53782011
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For sampling powders, granulates and fluids. Buerkle™ SteriPlast™ Blue Detectable Spoons are sterilized by gamma rays.

  • Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
  • This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
  • Can be used as part of HACCP/IFS foreign object management
  • Individually packaged for disposable use
  • Production, assembly and packaging according to clean room class 7
  • Can be detected by colour scanners, metal detectors or X-rays

Compliance

EU food requirements

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2.5 mL
Color Blue
Disposable Single-use
Product Type Detectable Sampling Spoon
For Use With (Application) Sampling Powders, Granulates and Fluids
Length (English) 5 in.
Material Polystyrene
Length (Metric) 127 mm
Sterility Sterile
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.