missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
- This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged for disposable use
- Production, assembly and packaging according to clean room class 7
- Can be detected by colour scanners, metal detectors or X-rays
Compliance
EU food requirements
Specifications
Specifications
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 10 mL |
| Color | Blue |
| Disposable | Single-use |
| Product Type | Detectable Sampling Spoon |
| For Use With (Application) | Sampling Powders, Granulates and Fluids |
| Length (English) | 6.6 in. |
| Material | Polystyrene |
| Length (Metric) | 170 mm |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?
