missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™/LaboPlast™Bio Palette Knife Spatula
BIOPHARMAGREENER_CHOICE

Product Code. 17250624 Shop All Bürkle Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 17250624

Brand: Buerkle™ 53791035

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle's dosing spoons.

  • The dimensionally stable long edge of the plastic scraper is ideal for cutting viscous media.
  • A small tip on the long edge can be used for ripping, for example packaging film.
  • The scraper is also suitable for stirring and blending.
  • Bio-PE, white, flexible and break-proof
  • Product made from renewable raw materials
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100
Sterility Sterile
Autoclavable Not Autoclavable
Certifications/Compliance Complies with EU food rrequirements and FDA regulations
Disposable Single-use
Product Type Sampler
Length (Metric) Blade 70 mm
Width (Metric) 20 mm
Material Bio-PE
Length (Metric) 192 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.