missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Spoons for Foodstuffs
BIOPHARMA

Product Code. 15694124 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
2.5 mL
10 mL
Length (Metric):
127 mm
170 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Unit Size
15694124 2.5 mL 127 mm Pack of 100
15604134 10 mL 170 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15694124 Supplier Buerkle™ Supplier No. 53783011
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For sampling powders, granulates, pastes and fluids. Buerkle™ SteriPlast™ Spoons for Foodstuffs are sterilized by gamma rays.

  • Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
  • This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
  • Can be used as part of HACCP/IFS foreign object management
  • Individually packaged for disposable use
  • Production, assembly and packaging according to clean room class 7
  • Can be detected by colour scanners

Compliance

EU food requirements

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2.5 mL
Color Blue
Disposable Single-use
Product Type Food Sampling Spoon
For Use With (Application) Sampling Powders, Granulates, Pastes and Fluids
Length (English) 5 in.
Material Polystyrene
Length (Metric) 127 mm
Sterility Sterile
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.