missing translation for 'onlineSavingsMsg'
Learn More

Merck Steritop™ E-GP Funnel-less Sterile Vacuum Filtration Systems

Product Code. 16467545
Change view
Click to view available options
Outlet Fitting:
38 mm threaded
45 mm threaded
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Outlet Fitting Unit Size
16467545 45 mm threaded Pack of 12
16457545 38 mm threaded Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16467545 Supplier Merck Supplier No. SEGPT0045
  • ONLINE EXCLUSIVE PRICE
    170.00 €
    152.99 € / Pack of 12
    Save 17.01 €
    10% Off
View all promotions
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.

Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.

The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers.

SEGPT0038 is a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane that can directly connect to any volume, 45 mm receiver flask (glass or plastic) for sterilizing tissue culture media and buffers.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear with blue adapter
Disposable Single-use
Material Polystyrene
Material (Membrane) PES Express PLUS
Pore Size 0.22 μm
Sterility Sterile
Filtration Area 40 cm²
For Use With (Application) Sterile filtration
Outlet Fitting 45 mm threaded
Outlet Style 45 mm threaded
Type Bottle Top Filter
Volume (Metric) <2 mL (limited by vial volume)
Diameter (Metric) 73 mm
Diameter (Metric) Filter 73 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.