Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
44.25 € - 261.02 €
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
16269421
|
Sampling Systems™
8370H-100S |
ItemDescription: Yes |
131.00 €
Pack of 50 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15504004
|
Sampling Systems™
8370H-200S |
ItemDescription: Yes |
121.00 €
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
16259421
|
Sampling Systems™
8370H-100 |
ItemDescription: No |
56.60 €
Pack of 50 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
15879741
|
Sampling Systems™
8370H-100 |
ItemDescription: No |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15360044
|
Sampling Systems™
8370H-100S |
ItemDescription: Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15889741
|
Sampling Systems™
8370H-200 |
ItemDescription: No |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Constructed of HDPE
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required