Learn More
Sampling Systems™ SteriWare™ Disposable Sack Sampler
Product Code. 11905856 Shop All Sampling Systems Products
Change view
Click to view available options
Width (Metric):
14 mm
25 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Sterility unitSize
11905856 25 mm Sterile Pack of 100
11995846 25 mm Non-sterile Pack of 100
15691582 14 mm Sterile Pack of 100
15839461 14 mm Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11905856 Supplier Sampling Systems™ Supplier No. 8085A100S
Add to basket
Add to basket
Sampling Systems™ SteriWare™ Disposable Sack Sampler is a hygienic, single use device that has been specially designed to take samples through the side of the sack.
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Suitable for use in ATEX areas
  • Available pre-sterilized (SAL 10-6)
Specifications
Material HDPE
Sterility Sterile
Length (Metric) 400 mm
Width (Metric) 25 mm