missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ HDPE Disposable Slot Sampler
BIOPHARMA

Product Code. 15210749 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
100 mL
500 mL
Unit Size:
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15210749 500 mL Pack of 20
12702467 100 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15210749 Supplier Sampling Systems™ Supplier No. 8073J1000S
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for taking a multilevel, cross sectional sample from powders and granules. The disposable Slot Sampler takes a single composite from the top, middle and bottom of your powder bed.

Key Features:

  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Available in 2 different lengths

Operation:

  • Push the sampler into the product. Ensuring the sampler is in the closed position.
  • Rotate handle to open up the slots.
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the ampler.
  • Remove sampler from the product.
  • Slide the inner rod out of the outer tube.
  • Deposit the sample into a suitable container.

Why Use a SteriWare™ Disposable Sampler?

  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.

Specifications

Product Type Multilevel Slot Sampler
Material HDPE
Capacity (Metric) 500 mL
Disposable Single-use
Sterility Sterile
Certifications/Compliance Conforms to FDA CFR 177.1520, EU Regulations 10/2011, EC Regulations 1935/2004
For Use With (Application) Ideal for taking fast multilevel, cross sectional samples from powders and granules
Length (Metric) 986 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.