Learn More
Sampling Systems™ SteriWare™ Open V Spatulas
BIOPHARMA
Product Code. 17524676 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric) Blade:
100 mm
150 mm
250 mm
Width (Metric) Blade:
10 mm
20 mm
Size:
Narrow
Standard
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Width (Metric) Blade Size Sterility Unit Size
17524676 - - - - -
17564666 100 mm 20 mm Standard Non-sterile Pack of 100
17574666 100 mm 20 mm Standard Sterile Pack of 100
17584666 150 mm 20 mm Standard Non-sterile Pack of 100
17594666 150 mm 20 mm Standard Sterile Pack of 100
17504676 250 mm 20 mm Standard Non-sterile Pack of 100
17514676 250 mm 20 mm Standard Sterile Pack of 100
17534676 100 mm 10 mm Narrow Sterile Pack of 100
17544676 150 mm 10 mm Narrow Non-sterile Pack of 100
17554676 250 mm 10 mm Narrow Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17524676 Supplier Sampling Systems™ Supplier No. 8440N100
  • ONLINE EXCLUSIVE PRICE
    113.00 €
    111.71 € / Pack of 100
    Save 1.29 €
    1% Off
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Improve your quality control by taking fixed volume powder samples easily with SteriWare™ Open V Spatulas. The open tip design is ideal for dispensing and sampling small quantities of powders. Available sterile in a wide range of sizes ideal for the pharmaceutical, chemical and food industry.
  • Our open ended disposable V spatula are ideal for sampling, raw material testing and general goods in testing.
  • Manufactured to the highest standard in a medical cleanroom to ensure cleanliness and supplied with full documentation.
  • Available in 2 widths and 3 lengths to ensure you have the right sampling tool for the job.
  • The narrow versions are ideal if you want to put the sample into the test tube or take a small sample.
  • The longer version is ideal when you want to take more of a cross sectional sample.
  • The disposable V spatula comes pre bagged and ready to use.
  • Simply unwrap and use.
  • No cleaning or cleaning validation is required.
  • Available in 2 versions, either sterile or non sterile versions.
  • Sterile and non sterile spatulas are ideal when a robust tool is required.
  • Made from high quality PS that conforms to the latest food grade, EC and FDA standards.
  • BSE/TSE free.
  • Batch certificates of conformity supplied.
  • Moulded and packed in a medical cleanroom to maintain cleanliness.