Learn More
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™
BIOPHARMAGREENER_CHOICE
Product Code. 12917945 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2.5 L
Unit Size:
Pack of 100
Pack of 15
Pack of 25
Pack of 50
Pack of 85
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12917945 60 mL Pack of 100
17488873 2.5 L Pack of 15
15510904 30 mL Pack of 100
17418883 500 mL Pack of 50
17478873 1000 mL Pack of 25
15819441 125 mL Pack of 100
17498873 250 mL Pack of 85
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12917945 Supplier Sampling Systems™ Supplier No. 8045R60S
Only null left
Add to basket
Add to basket
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Sterility Sterile
Capacity (Metric) 60 mL
Color Red
Disposable Single-use
Product Type Disposable Scoop
Includes Individually Bagged
Material Polystyrene