Learn More
Sampling Systems™ SteriWare Swabbing Templates
Product Code. 17972072 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Dimensions (L x W):
50 x 50 mm
100 x 100 mm
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Dimensions (L x W) unitSize
17972072 Sterile 50 x 50 mm Pack of 50
17992062 Non-sterile 50 x 50 mm Pack of 50
17942122 Non-sterile 100 x 100 mm Pack of 50
17982062 Sterile 100 x 100 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17972072 Supplier Sampling Systems™ Supplier No. 8888H50S
Add to basket
Add to basket
These swabbing templates are ideal for ensuring consistency when swabbing for cleaning validation purposes. They are made in a cleanroom and come individually bagged to help reduce the risk of contamination.
  • Available sterile or non-sterile
  • Ensures consistent validation results
Specifications
Product Type Swabbing Template
Material HDPE
Autoclavable Not Autoclavable
Disposable Single-use
Graduated No
Sterility Sterile
Dimensions (L x W) 50 x 50 mm