Learn More
STF 118804, Tocris Bioscience™
NAMPT Inhibitors
Brand:  Tocris Bioscience 5207/25
Encompass
Product Code. 15973211
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
STF 118804 is a NAMPT inhibitor (nicotinamide phosphoribosyltransferase, visfatin, PBEF1); inhibits NAD+ synthesis from nicotinamide. Potently inhibits the viability of multiple B-cell acute lymphoplastic leukemia (B-ALL) cell lines (IC50 < 10 nM); induces apoptosis in MF-411 cells. Induces regression of ALL xenografts and eliminates leukemia stems cells from bone marrow in mice.
Specifications
Specifications
4-[5-Methyl-4-[[(4-methylphenyl)sulfonyl]methyl]-2-oxazolyl]-N-(3-pyridinylmethyl)benzamide
894187-61-2
>98%
25 mg