Learn More
ibidi sticky-Slide Tissue
Product Code. 31941066
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
15/Cs.
90/Cs.
No. per Pack:
1 Ea.
8/Tray, 15 Trays
Unit Size:
Pack of 120
Pack of 15
Pack of 90
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case No. per Pack Unit Size
31941066 90/Cs. 1 Ea. Pack of 90
30130505 15/Cs. 1 Ea. Pack of 15
31941035 120/Cs. 8/Tray, 15 Trays Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31941066 Supplier ibidi Supplier No. 8051890
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A sticky channel slide designed for multiplexing immunofluorescence and immunohistochemistry stainings of tissue sections.
  • A self-adhesive chamber for customized cell-based assays and microscopy
  • Self-adhesive chamber for tissue sections and microscopy
  • Sterilized
  • Individually packed
Specifications
Type Chamber Slide
For Use With (Application) Cell-based Assays, Microscopy
No. per Case 90/Cs.
No. per Pack 1 Ea.
Sterility Sterile