Learn More
Lenz Laborglasintrumente Still Head for Steam Distillation
Product Code. 10622282
Change view
Click to view available options
Standard Taper Joint:
NS 29/32
Unit Size:
Each
Product Code. Standard Taper Joint unitSize
10622282 NS 29/32 Each
1 options
Product Code. 10622282 Supplier Lenz Laborglasintrumente Supplier No. 05033044
  • ONLINE EXCLUSIVE PRICE
    97.00 €
    63.16 € / Pack of 1
    Save 33.84 €
    34% Off
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With two cones NS 29/32 and one DIN thread GL 14
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Standard Taper Joint NS 29/32