Learn More
Eurofins DiscoverX STK33 Protein, Active
Description
STK33 (Serine/Threonine Kinase 33) is predicted to be involved in mutotic DNA damage checkpoint signaling and protein autophosphorylation. It also predicted to be located in perinuclear region of cytoplasm. It may play a specific role in the dynamic behavior of the intermediate filament cytoskeleton by phosphorylation of VIME (by similarity).
GenBank NM_030906
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 62.4 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥65% |
| Protein | STK33, active |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.