missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ STO-609 acetate
GREENER_CHOICE

Product Code. 18066052
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18066052 10 mg 10mg
18708087 50 mg 50mg
18739047 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18066052 Supplier Tocris Bioscience™ Supplier No. 1551/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective CaM kinase kinase inhibitor

STO-609 acetate is a selective, cell-permeable inhibitor of Ca2+-calmodulin-dependent protein kinase kinase (Ki values are 80 and 15 ng/ml for inhibition of CaM-KKα and CaM-KKβ respectively); competes for the ATP-binding site. Displays > 80-fold selectivity over CaMK1, CaMK2, CaMK4, MLCK, PKC, PKA and p42 MAPK.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid acetate
CAS 1173022-21-3
Assay Percent Range >98%
Quantity 10 mg
Solubility Soluble to 10mM with sonication in DMSO
Molecular Formula C19H10N2O3.C2H4O2
Formula Weight Observed MW: 374.35

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.