missing translation for 'onlineSavingsMsg'
Learn More

Stoddard Solvent Analytical, J.T. Baker™

Product Code. 15548834 Shop All J.T.Baker Products
Change view
Click to view available options
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15548834 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548834 Supplier JT Baker Supplier No. V11001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 8052-41-3
Quantity 20 L
Molecular Formula C8H15BrO2
UN Number 1268
InChI Key IMSPZLCTPSXORJ-UHFFFAOYSA-N
SMILES CC(=O)OCCCCCCBr
IUPAC Name 6-bromohexyl acetate
Molecular Weight (g/mol) 223.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.