missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STOX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00 € - 468.00 €
Specifications
| Antigen | STOX1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
STOX1 Polyclonal antibody specifically detects STOX1 in Human samples. It is validated for Western BlotSpecifications
| STOX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C10orf24, PEE4, storkhead box 1, storkhead-box protein 1, winged-helix domain-containing protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 520-800 of human STOX1 (NP_689922.3). KPSQTGPKEKPFQKPRSLDSSRIFDGKAKEPYAEQPNDKMEAESIYINDPTVKPINDDFRGHLFSHPQQSMLQNDGKCCPFMESMLRYEVYGGENEVIPEVLRKSHSHFDKLGETKQTPHSLPSRGASFSDRTPSACRLVDNTIHQFQNLGLLDYPVGVNPLRQAARQDKDSEELLRKGFVQDAETTSLENEQLSNDDQALYQNEVEDDDGACSSLYLEEDDISENDDLRQMLPGHSQYSFTGGSQGNHLGKQKVIERSLTEYNSTMERVESQVLKRNECY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 219736 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title