Learn More
Lenz Laborglasinstrumente™ Straight Adapter
Click to view available options
Standard Taper Joint:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Joint Type:
Cone
Socket
Unit Size:
Each
Description
- Straight glass olive
- Available in a range of joint sizes
Specifications
Specifications
| Type | Straight Adapter |
| Material | Duran Glass |
| Standard Taper Joint | NS 14/23 |
| Joint Type | Socket |