missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Straight Suction Tube for Buerkle™ ProfiSampler
Description
- Manufactured from either polyethylene or polyamide plastic
- Polyethylene tubes used for general sampling, polyamide tubes used for flammable liquids in VbF Hazard Category AI (electrically conducting)
- Diameter of 6 x 8 mm
- Not autoclavable
Specifications
Specifications
| Product Type | Suction Tube |
| Material | Polyethylene |
| Autoclavable | Not Autoclavable |
| Tube Material | Polyethylene |
| Style | Straight |
| Color | Translucent |
| For Use With (Application) | For General Sampling |
| Weight (Metric) | 0.8 kg |
| Diameter (Metric) Outer | 6 x 8 mm |
| Length (Metric) | 200 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?