Learn More
Riley™ Stream Evo Eco LED Safety Glasses
Product Code. 30274413
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
30274413 Polycarbonate Each
1 options
Product Code. 30274413 Supplier Riley™ Supplier No. RLY00390
  • ONLINE EXCLUSIVE PRICE
    24.92 €
    8.47 € / Pack of 1
    Save 16.45 €
    66% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The first to market safety eyewear without single use plastic. Stream Evo Eco provides all the protection, comfort and style as the Stream Evo but supplied in the microfiber pouch.
  • Stream Evo Eco Safety Glasses Provide Extreme Comfort and Wearability with a Range of Lens Options to Suit a Variety of Conditions
  • Single wraparound polycarbonate lens
  • Soft
  • Available in a smaller size - Stream Evo Small
  • No single use packaging
Specifications
Lens Coating Scratch-resistant, Anti-fog
Product Type Safety Glasses
Frame Color Gray
No. per Case 200/Cs.
Weight (English) 2.38 oz.
Weight (Metric) 67.5 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless Uni-lens
Autoclavable Not Autoclavable