missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity 50% suspension

Product Code. 30117602 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1000mL
20mL
4mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30117602 4 mL 4mL
30117603 20 mL 20mL
30117610 50 mL 50mL
30117606 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30117602 Supplier IBA Lifesciences Supplier No. 21250002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strep-Tactin™ 4Flow™ high capacity 50% Suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

  • Strep-Tactin™ 4Flow™ high capacity is a resin for purification of strep-tagged proteins via affinity chromatography
  • Product is optimized for column-based use and suitable for HPLC/FPLC
  • Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
  • 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
  • Targeted elution: no tedious titration steps required and multiple regeneration cycles possible
  • Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
  • Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock

Specifications

Binding Capacity 20 mg/mL resin
Capacity (Metric) 4 mL
Concentration 50% suspension
Description 4% agarose, crosslinked
Particle Size 50 to 150 μm
Form Resin Slurry
Quantity 4 mL
Protein Form Tetrameric protein
Target Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Type Bulk Resin
Final Product Type Affinity chromatography equipment
Ligand Type Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Stationary Phase Agarose
Sufficient For Resin sufficient for 2 times 1 mL gravity flow column
Format Bottle
Column Type Suspension
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.