missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity 50% suspension
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1000mL
20mL
4mL
50mL
Description
- Strep-Tactin™ 4Flow™ high capacity is a resin for purification of strep-tagged proteins via affinity chromatography
- Product is optimized for column-based use and suitable for HPLC/FPLC
- Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
- 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
- Targeted elution: no tedious titration steps required and multiple regeneration cycles possible
- Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
- Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications
Specifications
| Binding Capacity | 20 mg/mL resin |
| Capacity (Metric) | 20 mL |
| Concentration | 50% suspension |
| Description | 4% agarose, crosslinked |
| Particle Size | 50 to 150 μm |
| Form | Resin Slurry |
| Quantity | 20 mL |
| Protein Form | Tetrameric protein |
| Target | Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin |
| Type | Bulk Resin |
| Show More |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering