missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity 50% suspension

Product Code. 30117610 Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1000mL
20mL
4mL
50mL
Denne vare kan ikke returneres. Se returpolitik

Produktkode 30117610

missing translation for 'mfr': IBA Lifesciences 21250025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Strep-Tactin™ 4Flow™ high capacity 50% Suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

  • Strep-Tactin™ 4Flow™ high capacity is a resin for purification of strep-tagged proteins via affinity chromatography
  • Product is optimized for column-based use and suitable for HPLC/FPLC
  • Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
  • 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
  • Targeted elution: no tedious titration steps required and multiple regeneration cycles possible
  • Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
  • Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock

Tekniske data

Binding Capacity 20 mg/mL resin
Capacity (Metric) 50 mL
Concentration 50% suspension
Description 4% agarose, crosslinked
Particle Size 50 to 150 μm
Form Resin Slurry
Quantity 50 mL
Protein Form Tetrameric protein
Target Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Type Bulk Resin
Final Product Type Affinity chromatography equipment
Ligand Type Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Stationary Phase Agarose
Sufficient For Resin sufficient for 25 times 1 mL gravity flow column
Format Bottle
Column Type Suspension
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.