missing translation for 'onlineSavingsMsg'
Learn More

IBA GmbH™ Strep-Tactin™ Chromeo 488 Conjugate

Product Code. 15382006 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15382006 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15382006 Supplier IBA GmbH™ Supplier No. 21542050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IBA GmbH™ Strep-Tactin™ Chromeo 488 Conjugate is for detection of Strep-tag™ and Twin-Strep-tag™ fusion proteins.

  • For the detection of Strep-tag™ II and Twin-Strep-tag™ in Immunofluorescence, FACS and more
  • Contains Strep-Tactin™ conjugated to Chromeo 488
  • 1 x 50μg fluorescent conjugate

Specifications

Quantity 50 μg
Sample Volume 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.