Learn More
IBA Lifesciences Strep-Tactin™ Conjugate
Product Code. 12698247 Shop All IBA Lifesciences Products
Change view
Click to view available options
Product Type:
AP Conjugate
HRP Conjugate
Unit Size:
500µL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12698247 HRP Conjugate Each
12806516 AP Conjugate 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12698247 Supplier IBA Lifesciences Supplier No. 21502001
  • ONLINE EXCLUSIVE PRICE
    225.00 €
    165.00 € / Pack of 1
    Save 60.00 €
    26% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Strep-Tactin™ conjugates with AP or HRP for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins
  • The streptavidin variant Strep-Tactin™ is available as conjugate with horseradish peroxidase (HRP) or with alkaline phosphatase (AP) for fast and directed detection of N-terminally, C-terminally, or internally tagged Strep-tag™II or Twin-Strep-tag™ fusion proteins transferred to a blotting membrane or captured on a microplate.
  • Both conjugates are suitable for chromogenic reaction, whereas Strep-Tactin™ HRP is applicable for chemiluminescence reaction as well.
  • Depending on the host organism, protein extracts may contain small amounts of biotinylated proteins, e.g., the biotin carboxyl carrier protein (BCCP, 22 kDa) of E. coli.
  • Biotin Blocking Buffer is recommended for blocking of such biotinylated proteins on western blots prior to detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins with Strep-Tactin™ HRP to prevent the detection of false positive bands.
  • For this purpose, Biotin Blocking Buffer contains the tetrameric biotin binding protein avidin, which selectively binds biotinylated proteins.
Specifications
Content And Storage 4°C
Product Type HRP Conjugate