missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ Conjugate

Strep-Tactin™ conjugates with AP or HRP for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins

Brand:  IBA Lifesciences 2-1503-001

Code : LE

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12806516

  • 164.00 € / 500µL
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

  • The streptavidin variant Strep-Tactin™ is available as conjugate with horseradish peroxidase (HRP) or with alkaline phosphatase (AP) for fast and directed detection of N-terminally, C-terminally, or internally tagged Strep-tag™II or Twin-Strep-tag™ fusion proteins transferred to a blotting membrane or captured on a microplate.
  • Both conjugates are suitable for chromogenic reaction, whereas Strep-Tactin™ HRP is applicable for chemiluminescence reaction as well.
  • Depending on the host organism, protein extracts may contain small amounts of biotinylated proteins, e.g., the biotin carboxyl carrier protein (BCCP, 22 kDa) of E. coli.
  • Biotin Blocking Buffer is recommended for blocking of such biotinylated proteins on western blots prior to detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins with Strep-Tactin™ HRP to prevent the detection of false positive bands.
  • For this purpose, Biotin Blocking Buffer contains the tetrameric biotin binding protein avidin, which selectively binds biotinylated proteins.
Specifications

Specifications

0.5 mL
2°C to 8°C
AP Conjugate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.