Learn More
IBA Lifesciences Strep-Tactin™ PE or APC
Tienda IBA Lifesciences Productos
Click to view available options
Label or Dye:
APC Conjugate
PE Conjugate
Tamaño de la unidad:
50µL
Descripción
- Strep-Tactin™ conjugated to either R-Phycoerythrin (PE) or Allophycocyanin (APC) is suitable for staining or isolation of antigen-specific cytotoxic T cells (MHC I Streptamer™) or surface marker-specific cells (Fab-TACS™/Nano-TACS™) based on IBAs Strep-tag™ technology.
- To target specific cells, MHC I complexes, low affinity Fab fragments or nanobodies fused to a Twin-Strep-tag™ (MHC I-Streps, Fab-Streps or Nano-Streps) are required in addition to fluorescent Strep-Tactin™.
- Strep-Tactin™ multimerizes MHC I-Streps or Fab-/Nano-Streps, ensuring stable binding to cells.
- The resulting fluorescent complexes can be used to stain cells that are specific for the respective peptide-loaded MHC I molecule or for a selected surface marker.
- The addition of biotin reverses the multimerization and causes the dissociation of MHC I-Streps or Fab-/Nano-Streps from the cell surface.
- The complete removal of all labeling reagents preserves the cells’ full biological activity.
Especificaciones
Especificaciones
| Kit Contents | 50 tests a 1 to 2 x106 cells |
| Quantity | 50 Tests |
| Label or Dye | APC Conjugate |
For Research Use Only.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido