missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin

Product Code. 16486149 Shop All Cytiva Products
Change view
Click to view available options
Packaging Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
16486149 10 mL 10mL
16496149 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486149 Supplier Cytiva Supplier No. 29401324
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Chromatography resin for purification of recombinant proteins tagged with a Strep-tag™ II and Twin-Strep-tag™.

  • Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration
  • Compatible with a wide range of detergents, denaturants, and other additives
TRUSTED_SUSTAINABILITY

Specifications

Stability Stable in all commonly used aqueous buffers
For Use With (Application) Purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag
Storage Requirements 2 to 8°C in 20% ethanol
Binding Capacity Approx. 10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin
Particle Size 34 μm
Matrix Rigid, cross-linked agarose, spherical
pH Range 6 to 10 pH
Density (Metric) Approx. 5 mg/mL resin
Ligand Type Strep-Tactin XT
Packaging Quantity 10 mL
compliance-icons
Product Identifier
  • StrepTactin XT Sepharose, 10 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.