missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin

Chromatography resin for purification of recombinant proteins tagged with a Strep-tag™ II and Twin-Strep-tag™.

Brand:  Cytiva 29401326

Additional Details : CAS Number : 64-17-5 Weight : 0.05000kg


 View more versions of this product

Product Code. 16496149

  • 4258.00 € / 50mL
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration
  • Compatible with a wide range of detergents, denaturants, and other additives
Specifications

Specifications

Purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag
2 to 8°C in 20% ethanol
34 μm
6 to 10 pH
Strep-Tactin XT
Stable in all commonly used aqueous buffers
Approx. 10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin
Rigid, cross-linked agarose, spherical
Approx. 5 mg/mL resin
50 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • StrepTactin XT Sepharose, 50 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers