missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™XT Superflow™ High Capacity Column

Product Code. 16401485 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
5 x 0.2 mL
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
16401485 10 mL Each
16491475 5 x 0.2 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16401485 Supplier IBA Lifesciences Supplier No. 24034001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Strep-Tactin™XT Superflow™ high capacity column is a gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

  • Strep-Tactin™XT Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • They enable the addition of denaturing agents such as urea during the purification procedure.
  • In comparison to Strep-Tactin™XT Superflow™, Strep-Tactin™XT Superflow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 3- to 5-fold higher yield of purified protein.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.

Specifications

Concentration 6% agarose, crosslinked
Formulation Ready-to-use column with Strep-Tactin™XT high capacity resin
Content And Storage 2°C to 8°C
For Use With (Application) Purification of Strep-Tag™II and Twin-Strep-Tag™ Fusion Proteins
Type Pre-packed Column
Capacity (Metric) 10 mL

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.