Learn More
Description
It is available coupled to fluorescent dyes (DY-488, DY-549, DY-649).
The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm
Please note: For capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases, the use of StrepMAB-Immo or Strep-Tactin™XT is recommended due to the binding affinity in the pM range.
Specifications
Specifications
| Description | Not lyophilized |
| Kit Contents | Suspension in PBS |
| Content And Storage | Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C |
| Form | Liquid Suspension |
| Quantity | 75 μg |
| For Use With (Application) | RUO |
| Target | Strep-tag™II and Twin-Strep-tag™ |
| Type | Conjugate |
| Label or Dye | HRP Conjugate |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
