Learn More
IBA GmbH™ StrepMAB-Immo Chromeo 488 Conjugate
IBA GmbH™ StrepMAB-Immo Chromeo 488 Conjugate is and antibody for detection of Strep-tag™ and Twin-Strep-tag™ fusion proteins.
Brand: IBA GmbH™ 2-1546-050
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- For the detection of Strep-tag™ II and Twin-Strep-tag™ in Immunofluorescence, FACS and more
- Contains StrepMAB-Immo conjugated to Chromeo 488
- 1 x 50µg fluorescent conjugate
Specifications
50 μg |