Learn More
IBA Lifesciences Streptavidin

Handla allt IBA Lifesciences Produkter


Beskrivning
The streptavidin:biotin system is widely used for immobilization and detection of biotinylated molecules, like proteins and nucleic acids. IBA Lifesciences provides bulk amounts of lyophilized streptavidin in reliable and high quality at competitive prices to generate streptavidin-coated surfaces (microplates, SPR chips, beads) or detection reagents conjugated with, e.g., fluorescent dyes.
Please note that streptavidin is not applicable for detection, immobilization, and purification of Strep-tag™II and Twin-Strep-tag™ fusion proteins, since the binding affinity for both tags is too low.
Specifikationer
Specifikationer
| Color | White |
| Concentration | Lyophilized from a 25 mg/ mL solution |
| Particle Size | 13.3 kDa/subunit |
| Quantity | 1 g |
| Source | Recombinant, expression in E. coli |
| Form | Powder |
| Purity or Quality Grade | >95 % |
| Host Species | Streptomyces avidinii |
| Product Type | Tetrameric Protein |
For Research Use Only.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.