missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, solid

Product Code. 11570746
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11570746 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11570746 Supplier Invitrogen™ Supplier No. S888
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

  • A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
  • Binding activity of streptavidin is determined to be >11 units per mg protein
  • One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
  • Store at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Unconjugated
Quantity 5 mg
Content And Storage Storage Upon Receipt:≤-20°C, desiccate
Shipping Condition Room Temperature
Form Solid
Product Type Streptavidin Conjugate (fluorescent)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.