missing translation for 'onlineSavingsMsg'
Learn More

Abcam Streptavidin Beads 6FF

Product Code. 32119936
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119936 25 mL 25mL
32119223 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32119936 Supplier Abcam Supplier No. ab31909725mL
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable to bind biotin and biotinylated proteins.

Supplemental Information

  • Ligand: Streptavidin
  • Basic Beads: 6% Cross-linked agarose beads
  • Particle size: 45μm - 165μm
  • Storage Buffer: 80% PBS, 20% ethanol
  • pH Stability Range: 4-9
  • Maximum pressure: 0.3 MPa
  • Binding Capacity: >20 nmol Free Biotin/ml
.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Quantity 25 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.