missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin-CY5 (ZyMax Grade)
Click to view available options
Quantity:
1 mL
Unit Size:
Each
Description
Applications: Western blotting, ELISA, Immunohistochemistry, ISH
ZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.
ZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | Cy5 |
| Quantity | 1 mL |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Form | Liquid |
| Product Line | Invitrogen |
| Product Type | Streptavidin Conjugate (fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction