missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Streptavidin-FITC

Product Code. 11459025 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459025 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459025 Supplier MP Biomedicals™ Supplier No. 08623411
  • ONLINE EXCLUSIVE PRICE
    468.00 €
    443.10 € / 1mg
    Save 24.90 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

used for the detection or localization of biotinylated biomolecules.

Specifications

Concentration 2 to 5 μg/mL
Conjugate FITC (Fluorescein)
Quantity 1 mg
Content And Storage 0°C to 5°C
Form Liquid
For Use With (Application) Immunoassay, used for the detection or localization of Biotinylated biomolecules
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.