Learn More
R&D Systems™ Streptavidin-Fluorescein
Product Code. 18401580
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
Product Code. Quantity Unit Size
18401580 30 μL 30µL
1 options
Product Code. 18401580 Supplier R&D Systems™ Supplier No. 48003014
  • ONLINE EXCLUSIVE PRICE
    153.46 €
    135.45 € / 30µL
    Save 18.01 €
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with apoptosis detection reagents and kits
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30 μL
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type Streptavidin-Fluorescein
Product Line R&D Systems