missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Streptavidin HP SpinTrap™ Buffer Kit

Product Code. 10300795 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
16 Reactions
Unit Size:
1 set
Product Code. Quantity unitSize
10300795 16 Reactions 1 set
1 options
This item is not returnable. View return policy

Product Code. 10300795

Brand: Cytiva 28913568

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

10X binding/washing buffer (TBS), 10X elution buffer, and blocking buffer

Pre-made buffers optimized for small-scale affinity preparation and protein enrichment using the Streptavidin HP SpinTrap columns

  • Stock solutions can be diluted directly into the provided bottles for storage
  • Eliminates time-consuming buffer preparation, allowing fast, reproducible protein enrichment

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Streptavidin Sepharose HP, spin column and 96-well filter plates filter plates, 10X binding, Wash Buffer (TBS), 10X Elution Buffer, Blocking Buffer
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C
Particle Size 34 μm
Quantity 16 Reactions
Type SpinTrap Buffer Kit
Ligand Type Streptavidin
Sufficient For 16 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.